Assay Details
Gene Symbol: acl-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072243.9 | NP_504644.1 | 2 - 3 | 536 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
