Assay Details
Gene Symbol: ppt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072283.6 | NP_504684.2 | 4 - 5 | 641 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
