Assay Details
Gene Symbol: C13A2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072453.5 | NP_504854.4 | 4 - 5 | 722 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
