Assay Details
Gene Symbol: F40A3.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072630.6 | NP_505031.1 | 2 - 3 | 437 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: F40A3.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072630.6 | NP_505031.1 | 2 - 3 | 437 | 87 |
Set Membership: |
![]() ![]() ![]() |