Assay Details
Gene Symbol: grd-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307786.2 | NP_001294715.1 | 3 - 4 | 412 | 58 | |
NM_001368461.2 | NP_001355526.1 | 3 - 4 | 534 | 58 | ||
NM_001368462.3 | NP_001355524.1 | 3 - 4 | 534 | 58 | ||
NM_001368463.3 | NP_001355525.1 | 4 | 550 | 58 | ||
NM_001368464.3 | NP_001355528.1 | 3 | 361 | 58 | ||
NM_001368465.2 | NP_001355527.1 | 3 - 4 | 412 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
