Assay Details
Gene Symbol: acl-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383368.1 | 4 - 5 | 743 | 110 | ||
NM_073011.8 | NP_505412.2 | 4 - 5 | 780 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
