Assay Details
Gene Symbol: acl-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383368.1 | 6 - 7 | 1071 | 140 | ||
NM_073011.8 | NP_505412.2 | 6 - 7 | 1108 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
