Assay Details
Gene Symbol: lipl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073126.4 | NP_505527.1 | 4 - 5 | 700 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
