Assay Details
Gene Symbol: acl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269246.4 | NP_001256175.1 | 2 - 3 | 311 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
