Assay Details
Gene Symbol: acl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269246.4 | NP_001256175.1 | 4 - 5 | 719 | 62 | |
NM_001269247.3 | NP_001256176.1 | 1 - 2 | 262 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
