Assay Details
Gene Symbol: txt-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073300.4 | NP_505701.2 | 1 - 2 | 257 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: txt-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073300.4 | NP_505701.2 | 1 - 2 | 257 | 107 |
Set Membership: |
![]() ![]() ![]() ![]() |