Assay Details
Gene Symbol: nrx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129396.5 | NP_001122868.2 | 5 - 6 | 621 | 67 | |
NM_001269333.4 | NP_001256262.1 | 5 - 6 | 624 | 67 | ||
NM_001276930.3 | NP_001263859.1 | 5 - 6 | 611 | 67 | ||
NM_001276931.3 | NP_001263860.1 | 5 - 6 | 611 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
