Assay Details
Gene Symbol: lipl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073828.4 | NP_506229.1 | 2 - 3 | 251 | 162 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
