Assay Details
Gene Symbol: cest-1.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074105.4 | NP_506506.1 | 3 - 4 | 396 | 158 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
