Assay Details
Gene Symbol: hsp-90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074225.6 | NP_506626.1 | 4 - 5 | 1965 | 69 |
Gene Symbol: C47E8.11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383455.1 | 2 - 3 | 1669 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383455.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
