Assay Details
Gene Symbol: acs-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074468.6 | NP_506869.1 | 1 - 2 | 300 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
