Assay Details
Gene Symbol: sfxn-1.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029110.3 | NP_001024281.1 | 2 - 3 | 328 | 79 | |
NM_001029111.4 | NP_001024282.1 | 2 - 3 | 302 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
