Assay Details
Gene Symbol: cand-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074843.6 | NP_507244.1 | 5 - 6 | 1905 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
