Assay Details
Gene Symbol: Y116F11B.9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029047.2 | NP_001024218.1 | 1 - 2 | 107 | 157 | |
NR_002406.1 | 1 - 2 | 107 | 157 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
