Assay Details
Gene Symbol: grd-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075522.4 | NP_507923.2 | 12 - 13 | 2691 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
