Assay Details
Gene Symbol: grd-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075576.4 | NP_507977.2 | 4 - 5 | 1032 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
