Assay Details
Gene Symbol: acl-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075978.5 | NP_508379.1 | 1 - 2 | 194 | 93 | |
NM_075979.8 | NP_508380.2 | 1 - 2 | 197 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_075979.8 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
