Assay Details
Gene Symbol: acl-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075978.5 | NP_508379.1 | 3 - 4 | 543 | 113 | |
NM_075979.8 | NP_508380.2 | 3 - 4 | 546 | 113 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_075979.8 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
