Assay Details
Gene Symbol: acl-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075978.5 | NP_508379.1 | 4 - 5 | 783 | 83 | |
NM_075979.8 | NP_508380.2 | 4 - 5 | 786 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
