Assay Details
Gene Symbol: rgl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129668.5 | NP_001123140.1 | 8 - 9 | 1649 | 85 | |
NM_076149.7 | NP_508550.1 | 7 - 8 | 1663 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
