Assay Details
Gene Symbol: ddr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076171.6 | NP_508572.1 | 11 - 12 | 1876 | 83 | |
NM_076172.8 | NP_508573.1 | 11 - 12 | 1882 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
