Assay Details
Gene Symbol: ddr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076171.6 | NP_508572.1 | 6 - 7 | 866 | 67 | |
NM_076172.8 | NP_508573.1 | 6 - 7 | 872 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
