Assay Details
Gene Symbol: mboa-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383582.1 | 2 - 3 | 235 | 73 | ||
GenBank mRNA | AB305047.1 | - | 2 - 3 | 227 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
