Assay Details
Gene Symbol: mboa-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383582.1 | 5 - 6 | 853 | 102 | ||
GenBank mRNA | AB305047.1 | - | 5 - 6 | 845 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
