Assay Details
Gene Symbol: F46H5.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076818.6 | NP_509219.1 | 12 - 13 | 1454 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
