Assay Details
Gene Symbol: F16F9.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077033.8 | NP_509434.2 | 3 - 4 | 335 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: F16F9.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077033.8 | NP_509434.2 | 3 - 4 | 335 | 117 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |