Assay Details
Gene Symbol: him-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029411.3 | NP_001024582.1 | 7 - 8 | 2042 | 118 | |
NM_001373449.3 | NP_001359724.1 | 7 | 1981 | 118 | ||
NM_001373450.2 | NP_001360633.1 | 6 - 7 | 1687 | 118 | ||
NM_001373451.2 | NP_001360634.1 | 6 - 7 | 1687 | 118 | ||
NM_077234.7 | NP_509635.2 | 7 - 8 | 1981 | 118 | ||
GenBank mRNA | AF074901.1 | - | 7 - 8 | 1981 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
