Assay Details
Gene Symbol: acl-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047817.4 | NP_001041282.1 | 2 - 3 | 237 | 73 | |
NM_001047818.4 | NP_001041283.1 | 1 - 2 | 65 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
