Assay Details
Gene Symbol: acl-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047817.4 | NP_001041282.1 | 4 - 5 | 465 | 92 | |
NM_001047818.4 | NP_001041283.1 | 3 - 4 | 293 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
