Assay Details
Gene Symbol: F57C7.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077373.5 | NP_509774.3 | 20 - 21 | 3407 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: F57C7.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077373.5 | NP_509774.3 | 20 - 21 | 3407 | 129 |
Set Membership: |
![]() ![]() ![]() ![]() |