Assay Details
Gene Symbol: rhgf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077390.10 | NP_509791.4 | 23 - 24 | 3557 | 156 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
