Assay Details
Gene Symbol: plc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029446.3 | NP_001024617.1 | 8 - 9 | 1584 | 75 | |
NM_001029448.6 | NP_001024619.1 | 8 - 9 | 1546 | 75 | ||
NM_001129669.4 | NP_001123141.2 | 41 - 42 | 6424 | 75 | ||
NM_001136454.3 | NP_001129926.1 | 8 - 9 | 1546 | 75 | ||
NM_001136455.4 | NP_001129927.1 | 8 - 9 | 1768 | 75 | ||
NM_001373489.2 | NP_001360637.1 | 41 - 42 | 6403 | 75 | ||
NM_001373491.2 | NP_001360639.1 | 41 - 42 | 6433 | 75 | ||
NM_001373492.2 | NP_001360635.1 | 41 - 42 | 6454 | 75 | ||
NM_001373493.2 | NP_001360636.1 | 42 - 43 | 6574 | 75 | ||
NM_001373494.2 | NP_001359720.1 | 42 - 43 | 6595 | 75 | ||
NM_001373495.2 | NP_001360638.1 | 42 - 43 | 6604 | 75 | ||
NM_077404.6 | NP_509805.3 | 8 - 9 | 1546 | 75 | ||
GenBank mRNA | AF044576.1 | - | 8 - 9 | 1606 | 75 | |
AM992255.1 | - | 8 - 9 | 1546 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
