Assay Details
Gene Symbol: hum-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373506.3 | NP_001360186.1 | 8 | 1622 | 69 | |
NM_001373507.3 | NP_001359540.1 | 9 | 1686 | 69 | ||
NM_001383647.1 | 8 - 9 | 1706 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383647.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
