Assay Details
Gene Symbol: T25C12.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077518.5 | NP_509919.2 | 28 - 29 | 5949 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: T25C12.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077518.5 | NP_509919.2 | 28 - 29 | 5949 | 125 |
Set Membership: |
![]() ![]() ![]() ![]() |