Assay Details
Gene Symbol: gcp-2.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077527.5 | NP_509928.3 | 14 - 15 | 2113 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
