Assay Details
Gene Symbol: lsd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077599.6 | NP_510000.2 | 4 - 5 | 880 | 157 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
