Assay Details
Gene Symbol: vab-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047256.5 | NP_001040721.1 | 17 | 10211 | 80 | |
NM_001377640.2 | NP_001364629.1 | 22 | 11021 | 80 | ||
NM_001377641.2 | NP_001364630.1 | 19 | 10667 | 80 | ||
NM_001377642.2 | NP_001364643.1 | 17 | 10118 | 80 | ||
NM_001377643.3 | NP_001364644.1 | 16 | 9931 | 80 | ||
NM_001377644.3 | NP_001364645.1 | 16 | 9869 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
