Assay Details
Gene Symbol: grd-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026660.5 | NP_001021831.1 | 3 - 4 | 413 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: grd-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026660.5 | NP_001021831.1 | 3 - 4 | 413 | 97 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |