Assay Details
Gene Symbol: cox-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379886.1 | NP_001367080.1 | 1 - 2 | 113 | 89 |
Gene Symbol: Y70G10A.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382976.1 | 1 - 2 | 107 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001382976.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
