Assay Details
Gene Symbol: cox-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379886.1 | NP_001367080.1 | 4 - 5 | 394 | 109 |
Gene Symbol: Y70G10A.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382976.1 | 4 - 5 | 390 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
