Assay Details
Gene Symbol: UCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003046.1 | NP_001003046.1 | 533 | 123 | ||
GenBank mRNA | AB046106.1 | - | 533 | 123 | ||
AF201376.1 | - | 104 | 123 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
