Assay Details
Gene Symbol: MCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003016.2 | NP_001003016.2 | 1 - 2 | 769 | 87 | |
GenBank mRNA | AB093582.2 | - | 1 - 2 | 796 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
