Assay Details
Gene Symbol: CYP1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008720.1 | NP_001008720.1 | 1 - 2 | 46 | 66 | |
XM_014109343.2 | XP_013964818.1 | 1 - 2 | 94 | 66 | ||
GenBank mRNA | AB094347.1 | - | 1 - 2 | 46 | 66 | |
AB094348.1 | - | 1 - 2 | 46 | 66 | ||
AB094349.1 | - | 1 - 2 | 46 | 66 | ||
AB159771.1 | - | 1 - 2 | 46 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001008720.1, XM_014109343.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
