Assay Details
Gene Symbol: CYP1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008720.1 | NP_001008720.1 | 4 - 5 | 1091 | 117 | |
XM_014109343.2 | XP_013964818.1 | 4 - 5 | 1154 | 117 | ||
GenBank mRNA | AB094347.1 | - | 4 - 5 | 1091 | 117 | |
AB094348.1 | - | 4 - 5 | 1091 | 117 | ||
AB094349.1 | - | 4 - 5 | 1091 | 117 | ||
AB159771.1 | - | 4 - 5 | 1091 | 117 | ||
HG965165.1 | - | 3 - 4 | 1029 | 117 | ||
HG965166.1 | - | 3 - 4 | 1029 | 117 | ||
HG965167.1 | - | 3 - 4 | 1029 | 117 | ||
HG974425.1 | - | 3 - 4 | 1029 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
