Assay Details
Gene Symbol: CYP1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008720.1 | NP_001008720.1 | 6 - 7 | 1308 | 61 | |
XM_014109343.2 | XP_013964818.1 | 6 - 7 | 1371 | 61 | ||
GenBank mRNA | AB094347.1 | - | 6 - 7 | 1308 | 61 | |
AB094348.1 | - | 6 - 7 | 1308 | 61 | ||
AB094349.1 | - | 6 - 7 | 1308 | 61 | ||
HG965166.1 | - | 5 - 6 | 1246 | 61 | ||
HG974425.1 | - | 5 - 6 | 1246 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
